missing translation for 'onlineSavingsMsg'
Learn More

Staurosporine, MP Biomedicals™

Product Code. 11490472 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490472 1 mg 1mg
11480472 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490472 Supplier MP Biomedicals Supplier No. 0219140091
  • ONLINE EXCLUSIVE PRICE
    936.00 EUR
    891.45 EUR / 1mg
    Save 44.55 EUR
    4% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Possesses inhibitory activity against fungi and yeasts. Staurosporine, MP Biomedicals is an antibiotic that is isolated from Streptomyces species. It is a potent inhibitor of phospholipid/Ca2+-dependent protein kinases and is used to study the involvement

Starosporin is used to study the involvement of protein kinases in signal transduction pathways. It arrests normal cells in the G1 checkpoint and is a potent inhibitor of Protein kinase C and other protein kinases in vitro and in cultured cells. It is an inducer of apoptosis in many mammalian cell types.

  • Potent inhibitor of phospolipid/calcium dependent and cyclic nucleotide dependent protein kinases
  • Useful as a tool for studies on protein phosphorylation in the regulation of cellular functions
  • Inhibits the upregulation of VEGF expression in tumor cells
  • Partially reverses MDR, sensitizing cells with MDR phenotype to cytotoxic agents
  • Inhibits Pgp phosphorylation (functional significance of Pgp phosphorylation is ill defined)

Chemical Identifiers

CAS 62996-74-1
Molecular Formula C28H26N4O3
Molecular Weight (g/mol) 466.541
InChI Key HKSZLNNOFSGOKW-ZYSRIHRCSA-N
Synonym Staurosporin, Antibiotic AM-2282
PubChem CID 49831000
SMILES CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC

Specifications

Type Staurosporine
Physical Form Powder
Melting Point 237°C
Color Off-White
Quantity 1 mg
Solubility Information May be reconstituted by dissolving the lyophilized reagent in DMSO (5mg/mL, clear, colorless solution), DMF, ethyl acetate (5mg/mL, clear colorless solution) or methanol or water.
Formula Weight 466.5
Percent Purity ≥98%
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.