missing translation for 'onlineSavingsMsg'
Learn More

Staurosporine, ≥98%, MP Biomedicals™

Product Code. 11452131 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452131 100 μg 100µg
11462131 500 μg 500µg
11472131 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452131 Supplier MP Biomedicals Supplier No. 0219480580
  • ONLINE EXCLUSIVE PRICE
    377.00 EUR
    359.10 EUR / 100µg
    Save 17.90 EUR
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 62996-74-1
Molecular Formula C28H26N4O3
Molecular Weight (g/mol) 466.541
InChI Key HKSZLNNOFSGOKW-ZYSRIHRCSA-N
Synonym Antibiotic AM-2282
PubChem CID 49831000
SMILES CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC

Specifications

Type Staurosporine
Physical Form Powder
Color White
Quantity 100 μg
Formula Weight 466.5
Percent Purity ≥98%
Chemical Name or Material Staurosporine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.