Learn More
R&L Enterprises™ Stand Rings with Bosshead
Product Code. 11729037 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Inner:
75 mm
100 mm
130 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
11729037 75 mm Each
10796945 100 mm Each
12996836 130 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11729037
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
Provides for ease of connection on ring stands.
  • Bosshead with plastic-headed clamping screw
  • Accepts rods from 8mm to 12.5mm in diameter
  • Dimensions given are internal diameter of ring x center to center distance from bosshead to ring
Specifications
Material Steel
Diameter (Metric) Inner 75 mm
Includes With Bosshead
For Use With (Application) For Rods