Learn More
Elma™ Starter Kit
Product Code. 17846126
Change view
Click to view available options
For Use With (Equipment):
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17846126 Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17846126 Supplier Elma™ Supplier No. 1124042
  • ONLINE EXCLUSIVE PRICE
    51.10 EUR
    39.17 EUR / Pack of 1
    Save 11.93 EUR
    23% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Modular basket system kit with Elma™ Pins for use with the Elmasonic P, Elmasonic Select, and EASY.
  • Includes: 3 of each Elma Pin (0741 and 0772), 2 of each Elma Pin (0509, 0158, 0767, 0613 and 0614, 0021R and 0001R), 2 of each silicone holder side comb and bottom comb
  • Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
Product Type Modular Basket System
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY