Learn More
Heidolph™ StarFish Split Support Rod
Product Code. 10556056 Shop All Heidolph Instruments Products
Change view
Click to view available options
Height (Metric):
650 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
10556056 650 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10556056 Supplier Heidolph™ Supplier No. 5058106000
  • ONLINE EXCLUSIVE PRICE
    205.00 EUR
    200.00 EUR / Pack of 1
    Save 5.00 EUR
    2% Off
Estimated Shipment: 23-12-2026
Augment your StarFish magnetic stirrer workstation with this supporting central rod. Heidolph™ StarFish Support Rod allows you to securely add a variety of components to your workstation/system.
  • Secure central support rod.
  • Accepts a variety of accessories.
TRUSTED_SUSTAINABILITY
Specifications
Description Split support rod for accessories.
For Use With (Equipment) Starfish Workstation, MR-Hei Magnetic Stirrers
Height (Metric) 650 mm