Learn More
Heidolph™ StarFish 5-Way Clamp
Product Code. 11731709 Shop All Heidolph Instruments Products
Change view
Click to view available options
Includes:
Silicone straps
Velcro straps
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
11731709 Velcro straps Each
11781659 Silicone straps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11731709 Supplier Heidolph™ Supplier No. 5058101000
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
Hold glassware firmly in place on your StarFish magnetic stirrer workstation. Heidolph™ StarFish 5-Way Clamp features five telescopic arms which can be extended and locked in place to suit your needs.
  • For MR HEI Magnetic Stirrers and StarFish Workstations.
  • Holds glassware of virtually any size.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MR-HEI Magnetic Stirrers
Holds Glassware
Includes Velcro straps
Type Clamp
Voltage 100/115/220/230 V