Learn More
Koettermann™ Standard Tall Cabinet, 900x1920x366mm
Product Code. 14865301
Change view
Click to view available options
No. of Doors:
2
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Doors Unit Size
14865301 2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 14865301 Supplier Koettermann™ Supplier No. 30100073
  • ONLINE EXCLUSIVE PRICE
    1275.00 EUR
    1020.00 EUR / Pack of 1
    Save 255.00 EUR
    20% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Koettermann™ Standard Tall Cabinet, 900x1920x366mm provides suitable storage to accommodate a variety of laboratory needs.
  • Tall storage cabinet
  • Two doors
  • Three shelves
  • Lockable
Specifications
Dimensions (D x W x H) 366 x 900 x 1920 mm
Includes Lock
No. of Doors 2
No. of Shelves 3
Product Type Tall Storage Cabinet