Learn More
Koettermann™ Standard Table, Heavy Loads, Wheels, 1800x750x900mm
Product Code. 14865391
Change view
Click to view available options
Includes: Lab SGL Worktop
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
14865391 Includes: Lab SGL Worktop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 14865391 Supplier Koettermann™ Supplier No. 50700028
  • ONLINE EXCLUSIVE PRICE
    2415.00 EUR
    2024.68 EUR / Pack of 1
    Save 390.32 EUR
    16% Off
Estimated Shipment: 19-03-2027
Koettermann™ Standard Table, Heavy Loads, Wheels, 1800x750x900mm is a mobile laboratory table with a durable worktop and is designed for heavier than normal weight bearing.
  • Heavy-duty mobile table
  • Lab SGL worktop
  • Standing height
  • One shelf
Specifications
Product Type Mobile Table
Depth (Metric) 750 mm
Description Heavy-Duty, Standing Height
Height (Metric) 900 mm
Includes Lab SGL Worktop
No. of Shelves 1
Width (Metric) 1800 mm