Learn More
Koettermann™ Standard Table, Heavy Loads, Wheels, 1200x600x750mm
Product Code. 15588555
Change view
Click to view available options
Includes: TopResist Worktop
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15588555 Includes: TopResist Worktop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15588555 Supplier Koettermann™ Supplier No. 50700009
  • ONLINE EXCLUSIVE PRICE
    2055.00 EUR
    1445.00 EUR / Pack of 1
    Save 610.00 EUR
    29% Off
Estimated Shipment: 01-04-2027
Koettermann™ Standard Table, Heavy Loads, Wheels, 1200x600x750mm is a mobile laboratory table with a durable worktop and is designed for heavier than normal weight bearing.
  • Heavy-duty mobile table
  • TopResist worktop
  • Steel shelf
Specifications
Product Type Mobile Table
Depth (Metric) 600 mm
Description Heavy-Duty
Height (Metric) 750 mm
Includes TopResist Worktop
No. of Shelves 1
Shelf Material Steel
Width (Metric) 1200 mm