Learn More
Electrothermal™ Standard Reaction Vessels
Used with Electrothermal™ RS600 Reaction Station and Omni OS6000 Series Reaction Stations
Brand:  Electrothermal™ 36630-52
Additional Details : Weight : 0.80000kg
Product Code. 12514236
  • 1375.00 EUR / Pack of 6
Estimated Shipment: 16-07-2024
Description
  • For use with Electrothermal™ RS600 Reaction Station and Omni OS6000 Series Reaction Stations.
  • 250mL
  • Round Bottomed Flasks
  • 56.5mm diameter
  • Quantity: 6/PK
Specifications
Specifications
For use with RS600 Reaction Station and Omni OS6000 Series Reaction Stations