Learn More
Thermo Scientific™ Manual, Removable-Needle, Gas Tight Syringes for GC Instruments
Product Code. 15274583 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
5 mL
10 mL
Needle Gauge:
22 G
22s G
23 G
25 G
26s G
Needle Point Style:
90° Blunt
Bevel
Blunt
Side Hole
Unit Size:
Each
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Needle Gauge Needle Point Style unitSize
15274583 10 mL 22s G Side Hole Each
10111033 10 μL 22 G 90° Blunt Each
10552455 25 μL 22s G Blunt Each
10630564 50 μL 22 G 90° Blunt Each
10523035 100 μL 22 G 90° Blunt Each
10394563 250 μL 22 G Blunt Each
10435923 500 μL 22s G Blunt Each
15234573 10 μL 26s G Bevel Each
11524070 1 mL 22s G Blunt Each
15224583 5 mL 22 G Bevel Each
15224603 5 mL 22s G Blunt Each
15244583 10 mL 22 G Bevel Each
15254583 10 mL 22s G Blunt Each
10117714 25 μL 25 G Bevel Each
10127744 500 μL 25 G Bevel Each
10640626 1 mL 23 G Bevel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 15274583 Supplier Thermo Scientific™ Supplier No. 365N8923
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
Use Thermo Scientific™ Removable Needle Syringes for accurate and reproducible manual sample introduction into GC instruments, with or without the needle.
Get durability, clarity and accuracy from Thermo Scientific™ Manual Removable Needle Syringes. Used for manual sample injection into gas chromatography (GC) instruments with or without the needle, they provide accuracy and reproducibility that provides confidence in analytical results, time-after-time. Choose from a variety of sizes, and types. These syringes are available in standard industry size with black easy-to-read and solvent-resistant markings.
Durability
The enhanced design of the syringes delivers:
  • Improved solvent resistance
  • Greater temperature range
  • Increased operational smoothness
  • Longer lifetime and improved cycle life
Clarity
Syringes are provided with a clear, easy-to-read black scale, which aids in consistent delivery of samples for manual syringes and easy identification of installed autosampler syringes.
Accuracy
The enhanced design of the syringe has eliminated areas where the sample can become trapped and potentially cause carryover:
  • Closer fit between the plunger tip and PTFE insert at the zero position
  • Tighter fit between PTFE insert and glass barrel
  • Improved fixed needle attachment
In addition, the reduction in adhesive from the fluid path enhances durability and reduces the risk of sample interaction with the adhesive.
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 22s G
Length (Metric) 50 mm
Product Type Gas Tight GC Autosampler Syringe, Removable Needle
Needle Point Number 365 RN461
Quantity 1/Ea.
Volume (Metric) 10 mL
Length (Metric) Needle 50 mm
Needle Point Style Side Hole
For Use With (Equipment) Gas Chromatography Systems
Replacement Needle 365RN465