Learn More
Koettermann™ Standard Lower Cabinet with Wheels, 900x740x516mm
Product Code. 15538515
Change view
Click to view available options
Two doors
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15538515 Two doors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15538515 Supplier Koettermann™ Supplier No. 31500136
  • ONLINE EXCLUSIVE PRICE
    842.00 EUR
    733.00 EUR / Pack of 1
    Save 109.00 EUR
    12% Off
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
Koettermann™ Standard Lower Cabinet with Wheels, 900x740x516mm provides suitable storage to accommodate a variety of laboratory needs.
  • Mobile cabinet
  • Two doors
  • One removable shelf
Specifications
Product Type Mobile Cabinet
Dimensions (D x W x H) 516 x 600 x 740 mm
Locking Mechanism No
No. of Doors 2
No. of Shelves 1