Learn More
Koettermann™ Standard Individual Table, C-Frame, 1500x750x750mm
Product Code. 15598545
Change view
Click to view available options
Includes: TopResist Worktop
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15598545 Includes: TopResist Worktop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15598545 Supplier Koettermann™ Supplier No. 50500057
  • ONLINE EXCLUSIVE PRICE
    1110.00 EUR
    909.00 EUR / Pack of 1
    Save 201.00 EUR
    18% Off
Estimated Shipment: 01-04-2027
Koettermann™ Standard Individual Table, C-Frame, 1500x750x750mm is a free-standing table with a durable worktop and C-type legs.
  • Table with C-type legs
  • TopResist worktop
Specifications
Product Type Table
Depth (Metric) 750 mm
Height (Metric) 750 mm
Includes TopResist Worktop
Mounting Type C-type legs
Width (Metric) 1500 mm