Learn More
Standard Buffer Solutions, WTW™
GREENER_CHOICE
Product Code. 11795204
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11795204 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11795204 Supplier WTW Supplier No. 109130
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
As a standard buffering solution, this buffer pH 9.180 (PL 7) is available in a 250 ml bottle.
  • Achieve accurate pH readings  
  • Available in convenient, ready-made pH value 9.18
  • Standard buffering solutions by WTW are traceably certified to PTB/NIST and therefore safe for use
  • Certificate of analysis from DIN/NIST
  • Standard buffering solution,with pH 9.180 and PL 7 that is available in a 250 ml bottle
  • Accurate to ± 0.02 pH
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 100°C
Color Colorless
Certificate of Analysis DIN/NIST
Density 0.999g/cm3
For Use With (Application) Precision applications
pH 9.18
Physical Form Liquid
Quantity 250 mL
Packaging Bottle
compliance-icons
Product Identifier
  • PL 9
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: Disodium tetraborate, decahydrate