Learn More
Buerkle™ Worm-Threaded Hose Clips
Product Code. 10595624 Shop All Bürkle Products
Change view
Click to view available options
Tubing Size:
10 to 16 mm dia.
12 to 20 mm dia.
16 to 27 mm dia.
20 to 32 mm dia.
25 to 40 mm
8 to 12 mm dia.
Unit Size:
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size unitSize
10595624 8 to 12 mm dia. Pack of 10
16616703 25 to 40 mm Pack of 10
15784068 20 to 32 mm dia. Pack of 10
10428151 10 to 16 mm dia. Pack of 10
10263020 12 to 20 mm dia. Pack of 10
10550784 16 to 27 mm dia. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10595624 Supplier Buerkle™ Supplier No. 86770812
  • ONLINE EXCLUSIVE PRICE
    14.20 EUR
    13.95 EUR / Pack of 10
    Save 0.25 EUR
    1% Off
View all promotions
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Manufactured from stainless steel 1.4016.
  • With screw thread
  • Screw is zinc-coated
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 8 to 12 mm
Material Stainless Steel
For Use With Hoses
Packaging Quantity 10/Pk.
Tubing Size 8 to 12 mm dia.
Product Type Worm-threaded Hose Clip