Learn More
Bochem™ Stainless Steel Wire Basket, Square
Product Code. 10545031
Change view
Click to view available options
Dimensions (L x W x H):
150 x 150 x 150 mm
160 x 160 x 160 mm
180 x 180 x 180 mm
200 x 200 x 150 mm
200 x 200 x 200 mm
250 x 150 x 150 mm
300 x 200 x 150 mm
400 x 300 x 200 mm
Array:
160 mm
180 mm
200 mm
250 mm
300 mm
400 mm
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Array unitSize
10545031 200 x 200 x 150 mm 200 mm Each
10000751 150 x 150 x 150 mm - Each
10468812 180 x 180 x 180 mm 180 mm Each
10603462 160 x 160 x 160 mm 160 mm Each
10545221 250 x 150 x 150 mm 250 mm Each
10654032 300 x 200 x 150 mm 300 mm Each
10644032 200 x 200 x 200 mm 200 mm Each
10244271 400 x 300 x 200 mm 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10545031 Supplier Bochem™ Supplier No. 10030
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
Store laboratory items in the Bochem™ Stainless Steel Wire Basket, Square. It is sturdy and stable.
  • Stainless steel
  • Square
  • Flat bottom for stability
Specifications
Dimensions (L x W) 200 L x 200 Wmm
Dimensions (L x W x H) 200 x 200 x 150 mm
Product Type Wire Basket
Array 200 mm