Learn More
Tenak™ Stainless Steel Upright Freezer Rack for Cryogenic Boxes
Product Code. 15200664 Shop All TENAK Products
Change view
Click to view available options
Holds:
Boxes (16)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
15200664 Boxes (16) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15200664 Supplier Tenak™ Supplier No. TE23132B
  • ONLINE EXCLUSIVE PRICE
    164.00 EUR
    161.07 EUR / Pack of 1
    Save 2.93 EUR
    1% Off
Estimated Shipment: 09-02-2027
Use Tenak™ Stainless Steel Upright Freezer Racks for Cryogenic Boxes for your cryogenic storage needs. A variety of stainless steel racks accommodate box heights and quantities for laboratory freezers.
  • Suitable for all brands of upright freezers
  • Stainless steel
  • For cryogenic boxes of various capacities
Specifikationer
Cell Dividers Yes
Holds Boxes (16)
Material Stainless Steel
Positions 16
For Use With (Application) Fit into any freezer brand and model
For Use With (Equipment) Upright Freezers
No. per Pack 1
Quantity 1 Ea.
Depth (Metric) 558 mm
Product Type Standard Rack
Shelves 16
Height (Metric) 221 mm
Array 4 x 4
Width (Metric) 139 mm
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.