Description
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Stainless-Steel Unions, Tees and Crosses.
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ Stainless-Steel Unions, Tees and Crosses. Includes two stainless-steel nuts and ferrules.
- Absolute zero or low dead volume formats
- Includes two stainless steel nuts and ferrules
Specifications
Specifications
| Type | Union |
| Swept Volume | 0.025 μL |
| Includes | Union with 2 stainless steel nuts and ferrules |
| Description | 0.010 in. through hole |
| Material | Stainless Steel |
| For Use With (Equipment) | Upchurch Scientific/Parker fittings |