Learn More
R&L Enterprises™ Rectangular Retort Stand Bases
Product Code. 11705386 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric) Base:
160 mm
200 mm
250 mm
315 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Base Unit Size
11705386 250 mm Each
11779593 160 mm Each
11795376 200 mm Each
11715396 315 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11705386
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
An excellent option for supporting retort stands in the laboratory. R&L Enterprises™ Rectangular Retort Stand Bases are manufactured from white acrylic finished steel.
  • Without rod
  • Fitted with rubber feet
  • Hole threaded M10 at the center of one short side
Specifications
Material Stainless Steel
Length (Metric) Base 250 mm
Width (Metric) Base 160 mm
Includes With Rubber Feet
Product Type Tripod Plate
Color White