Learn More
UVP Stainless Steel TFML-40 Model Transilluminator
Product Code. 12873938
Change view
Click to view available options
Wavelength:
302/365 nm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength unitSize
12873938 302/365 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12873938 Supplier UVP Supplier No. 95042602
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type High Performance 2UV Transilluminator
Filtration Area (Metric) 200 x 400 mm
Electrical Requirements 230 V 50 Hz
Size (Metric) Filter 200 x 400 mm
Model TFML-40
Voltage 230 V
Frequency 50 Hz
Intensity Single
Wavelength 302/365 nm
Lighting Type 2UV Benchtop