Learn More
RETSCH Stainless Steel Collecting Pan for Test Sieves
Product Code. 11964422 Shop All Retsch Products
Change view
Click to view available options
Diameter (Metric) Outer:
100 mm
150 mm
200 mm
203 mm
305 mm
315 mm
400 mm
450 mm
Height (Metric):
25 mm
25.4 mm
40 mm
50 mm
50.8 mm
65 mm
75 mm
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Height (Metric) Unit Size
11964422 203 mm 25.4 mm Each
10360182 100 mm 40 mm Each
10146021 150 mm 40 mm Each
10153262 200 mm 25 mm Each
10136071 400 mm 65 mm Each
10790843 305 mm 40 mm Each
11347550 450 mm 50 mm Each
12549737 315 mm 75 mm Each
10238981 203 mm 50.8 mm Each
10585072 200 mm 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11964422 Supplier RETSCH Supplier No. 69.720.3025
  • ONLINE EXCLUSIVE PRICE
    96.80 EUR
    80.45 EUR / Pack of 1
    Save 16.35 EUR
    16% Off
Estimated Shipment: 22-02-2027
Collects resulting samples from test sieves. These Stainless Steel pans are compatible with multiple test sieves and sieve shakers, based upon the required diameter.
  • Stainless Steel
  • Compatible with multiple test sieves, based upon diameter
Specifications
Product Type Collecting Pan
Material Stainless Steel
For Use With (Equipment) Test Sieve
Height (Metric) 25.4 mm
Diameter (Metric) Outer 203 mm