missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Stainless Steel Temperature Probe

Product Code. 11507899
Change view
Click to view available options
Temperature (Metric) Range:
-5°C to 105deg;C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature (Metric) Range unitSize
11507899 -5°C to 105deg;C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11507899 Supplier Hanna Instruments™ Supplier No. HI7662T

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Allows for temperature compensated pH measurements on Hanna benchtop meters with a phono connector input. Hanna Instruments™ Stainless Steel Temperature Probe is an NTC thermistor with a 1m cable that features a wide measurement range.

pH readings must be temperature compensated in order to obtain accurate results. Temperature compensation provides the actual pH of the buffer or sample at the temperature of measurement. The HI7662-T is a separate high-accuracy temperature probe that can be used with Hanna benchtop meters and potentiometric titration systems.

  • Made of AISI 316 stainless steel
  • NTC thermistor temperature probe
  • Wide measurement range: -5.0 to 105.0°C
  • High speed to accommodate reactions whose temperature can change rapidly

Specifications

Accuracy (Temperature) ±0.1°C
Type NTC Thermistor Temperature Probe
Includes 1m Cable
Material Stainless Steel
Resolution (Temperature) 0.1°C
Temperature (Metric) Range -5°C to 105deg;C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.