Learn More
Branson Ultrasonics™ Stainless Steel Tanks ABS and Polystyrene Cart Cover Ultrasonic Bath
Product Code. 10049410
Change view
Click to view available options
Frequency:
40 kHz
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Frequency Unit Size
10049410 40 kHz Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10049410 Supplier Branson Ultrasonics™ Supplier No. 000001100
  • ONLINE EXCLUSIVE PRICE
    237.30 EUR
    177.00 EUR / Pack of 1
    Save 60.30 EUR
    25% Off
View all promotions
Estimated Shipment: 30-11-2026
Specifications
Frequency 40 kHz
Capacity (Metric) 0.5 L
Depth (Metric) Interior 90 mm
Depth (Metric) Tank 115 mm
Height (Metric) Exterior 112 mm
Height (Metric) Interior 55 mm
Wattage 40 w
Width (Metric) 225 mm
Width (Metric) Interior 115 mm