Learn More
Fisherbrand™ Stainless Steel Tank, 18L
Product Code. 17225556 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
18 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17225556 18 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17225556
Estimated Shipment: 18-03-2027
For water bath.
For use with Fisherbrand™ Digital Heating Circulator.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 18 L
Dimensions (D x W x H) 420 x 690 x 420 mm