Learn More
VACUUBRAND™ Stainless Steel T-Pieces With Small KF Flanges
Specifications
Specifications
| Diameter (Metric) Inside | 16/16/16 mm |
| For Use With | Solenoid-operated valves VV and VV C |
| Material | Stainless Steel |
| Quantity | 1 Each |
| Size | 40 x 80 mm |
| Type | T-piece |