Learn More
Karl Hecht™ Stainless Steel Square Wire Baskets
Product Code. 10669431
Change view
Click to view available options
Size:
16 x 16 x 16 cm
20 x 20 x 20 cm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
10669431 20 x 20 x 20 cm Each
10328731 16 x 16 x 16 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10669431 Supplier Karl Hecht™ Supplier No. 41075020
  • ONLINE EXCLUSIVE PRICE
    172.53 EUR
    148.56 EUR / Pack of 1
    Save 23.97 EUR
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 28-01-2027
An excellent option for general storage applications in the laboratory.
  • Wire mesh construction so contents are visible
  • Square shape
  • Available in a range of sizes
Specifications
Description Wire mesh construction
Material Stainless Steel
Shape Square
Size 20 x 20 x 20 cm
Product Type Wire Basket