Learn More
S Murray and Co™ Stainless Steel Spring Forceps
Product Code. 12740916
Change view
Click to view available options
Length (Metric):
115 mm
125 mm
150 mm
Tip Style:
Blunt, Narrow Flutes
Sharp, Curved, Narrow Flutes
Sharp, Narrow Flutes
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tip Style unitSize
12740916 150 mm Blunt, Narrow Flutes Each
12391369 125 mm Blunt, Narrow Flutes Each
12381369 125 mm Sharp, Curved, Narrow Flutes Each
11340705 115 mm Sharp, Narrow Flutes Each
12371369 125 mm Sharp, Narrow Flutes Each
12700046 150 mm Sharp, Narrow Flutes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 12740916 Supplier S Murray and Co™ Supplier No. E003/03
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Use these forceps for applications such as major and minor surgery, post mortem, dissecting and dressing sets. S Murray™ Stainless Steel Spring Forceps comply with the requirements of the Medical Devices Directive 93/42/EEC, with full traceability maintained throughout the manufacturing process.
Use these forceps for applications such as major and minor surgery, post mortem, dissecting and dressing sets. S Murray™ Stainless Steel Spring Forceps comply with the requirements of the Medical Devices Directive 93/42/EEC, with full traceability maintained throughout the manufacturing process.
  • Stainless Steel forceps for dissection
Specifications
Product Type Spring Forceps
Material Stainless Steel
For Use With (Application) For dissection
Quantity Each
Tip Style Blunt, Narrow Flutes
Length (Metric) 150 mm