Description
- Sturdy and robust spigot made of stainless steel AISI 304 (1.4301)
- Seals PTFE, FKM
- Self-closing
- Thread connection 3/4 in.
- Lockable
- NW 10 mm
Specifications
Specifications
| Color | Silver |
| For Use With (Equipment) | R 3/4 in. Thread Connection |
| Material | Stainless Steel (1.4301) |
| Product Type | Spigot |