Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN55plus, 15 mm
Product Code. 17508705 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SN55plus Sterilizer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17508705 SN55plus Sterilizer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17508705 Supplier Memmert™ Supplier No. SN55plusB02072
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Non-perforated slide-in drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) SN55plus Sterilizer
Product Type Drip Tray
Height (Metric) Tray Rim: 15 mm
Material Stainless Steel