Learn More
Fisherbrand™ Stainless Steel Upright Freezer Rack
Product Code. 11835390 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
16 Boxes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
11835390 16 Boxes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11835390
  • ONLINE EXCLUSIVE PRICE
    156.00 EUR
    116.93 EUR / Pack of 1
    Save 39.07 EUR
    25% Off
 Restricted Item
In Stock
Add to basket
Add to basket
Accommodates most major brands up to 53 cm tall. Stainless Steel Upright Freezer Rack provides safe storage for freezing samples.
  • Robust mounting brackets assure handles stay securely in place for easy use.
  • Racks store boxes up to a 135 x 135 mm footprint and accommodate boxes up to 53 mm tall.
Specifications
Color Silver
Height (Metric) 225 mm
Height (English) 8.9 in.
Length (Metric) 556 mm
Length (English) 21.9 in.
Width (Metric) 140 mm
Width (English) 5.5 in.
Dimensions (L x W x H) 556 x 140 x 225 mm
For Use With (Equipment) 53 mm high cryoboxes
Holds 16 Boxes
Quantity 1 Ea.
Array 4 x 4
Material Stainless Steel
Product Type Cryogenic Storage Rack
Withstand Temperature (Metric) -20°C