Learn More
Sampling Systems™ Stainless Steel Pointed Scissors
Product Code. 15879641 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
150 mm
200 mm
Length (Metric) Blade:
45 mm
100 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Length (Metric) Blade Unit Size
15879641 150 mm 45 mm Each
15889641 200 mm 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15879641 Supplier Sampling Systems™ Supplier No. A364150
  • ONLINE EXCLUSIVE PRICE
    5.85 EUR
    5.63 EUR / Pack of 1
    Save 0.22 EUR
    3% Off
View all promotions
Estimated Shipment: 12-01-2027
These stainless steel scissors are made from hygienic 420 grade stainless steel with a double point design.
  • Polished finish
  • Two sizes available
  • 420 grade stainless steel
Specifications
Length (Metric) 150 mm
Length (Metric) Blade 45 mm
Material Stainless Steel (AISI 420)
Tip Style Double Point