Learn More
Sherwood Scientific™ Stainless Steel Nebulizer Assembly
Product Code. 11891835
Change view
Click to view available options
Type:
Stainless Steel Nebulizer Assembly
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11891835 Stainless Steel Nebulizer Assembly Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11891835 Supplier Sherwood Scientific™ Supplier No. 42008102
  • ONLINE EXCLUSIVE PRICE
    762.00 EUR
    630.03 EUR / Pack of 1
    Save 131.97 EUR
    17% Off
Estimated Shipment: 12-11-2026
A nebulizer for use with Sherwood™ Flame photometers, models 420 and 425.
  • For aspiration of solutions
  • Aids in atomic spectroscopy
Specifications
For Use With (Equipment) Sherwood™ Flame Photometers, models 420 and 425
Type Stainless Steel Nebulizer Assembly