Learn More
Swann-Morton™ Stainless Steel Minor Skin Graft Blade
Product Code. 12363680
Change view
Click to view available options
Size:
# SG3
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12363680 # SG3 Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12363680 Supplier Swann-Morton™ Supplier No. 2201
  • ONLINE EXCLUSIVE PRICE
    13.45 EUR
    12.13 EUR / Pack of 20
    Save 1.32 EUR
    9% Off
View all promotions
 Restricted Item
Estimated Shipment: 29-12-2026
Order Info
Size
Specifications
Blade Material Stainless Steel
Product Type Minor Skin Graft Blade
For Use With (Application) Debridement of Burn related Wounds at the site of Major Accidents and Disasters
Size # SG3
For Use With (Equipment) Fits # 3 Graduated, # 3L, # 5B, # 7, # 9, # B3 and # B3L Handles
Sterility Sterile
Quantity 20/Pk.