Learn More
Bochem™ Stainless Steel Microscopy Scissors
Product Code. 10218610
Change view
Click to view available options
Length (Metric):
100 mm
115 mm
Blade Style:
Bent
Straight
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Blade Style unitSize
10218610 100 mm Bent Each
10065090 100 mm Straight Each
10112511 115 mm Straight Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10218610 Supplier Bochem™ Supplier No. 4071
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
Manufactured from stainless steel. Bochem™ Stainless Steel Microscopy Scissors come in three handle types.
  • Manufactured from stainless steel
  • Three types: round-round, sharp-round, sharp-sharp
Specifications
Product Type Microscopy Scissors
Blade Style Bent
Length (Metric) 100 mm
Material Stainless Steel
Tip Style Sharp/Sharp