Learn More
Fisherbrand™ Stainless Steel Micro Chattaway Spatulas
Product Code. 11523482 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
100 mm
130 mm
150 mm
200 mm
Unit Size:
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11523482 100 mm Pack of 5
11533482 130 mm Pack of 5
11543482 150 mm Pack of 5
11553482 200 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11523482
  • ONLINE EXCLUSIVE PRICE
    12.95 EUR
    10.91 EUR / Pack of 5
    Save 2.04 EUR
    15% Off
View all promotions
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
Scrapes and lifts materials from tiny bottles or flasks. Fisherbrand™ Stainless Steel Chattaway Spatulas are non-magnetic with a bright finish.
  • Manufactured from Stainless Steel
  • Bent and rounded end scrapes and lifts materials from bottles and flasks
  • Fits into tight flasks and bottles with necks ≥ 4mm diameter
Specifications
Quantity 5 Pk.
Product Type Chattaway Spatula
Width (Metric) 4 mm
Diameter (Metric) Outer 1.6 mm (Barrel)
Dimensions (L x W) 100 x 4 mm
Material Stainless Steel
Length (Metric) 100 mm