Learn More
Fisherbrand™ Stainless Steel Manual Decapper for clean room
Product Code. 11520535 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
13 mm
20 mm
32 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11520535 20 mm Each
11845722 - Each
15259968 32 mm Each
11586034 13 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11520535
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
Allow easy removal of aluminum seals without breakage
  • Stainless Steel
  • Sterilizable for cleanroom applications
  • Manual
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Type Manual Decapper
For Use With (Equipment) Aluminum Cap
Size 20 mm