Learn More
Amsta Rostfritt Stainless Steel Lid For 608/609-75
Product Code. 30284959
Change view
Click to view available options
Capacity (Metric):
None
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
30284959 None Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30284959 Supplier Amsta Rostfritt Supplier No. 5010400
  • ONLINE EXCLUSIVE PRICE
    75.15 EUR
    69.20 EUR / Pack of 1
    Save 5.95 EUR
    7% Off
Estimated Shipment: 14-12-2026
Highest Quality Stainless Steel lid For 608/609-75 Amsta transport container. Manufactured in Sweden.
  • Stainless steel 304 /1.4301
Specifications
Product Type Container Lid
Capacity (Metric) None
Color Silver
Material Stainless Steel