Learn More
Amsta Rostfritt Stainless Steel Lid - Acid Proof For 608/609-150
Product Code. 30284915
Change view
Click to view available options
Capacity (Metric):
None
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
30284915 None Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30284915 Supplier Amsta Rostfritt Supplier No. 5110550
  • ONLINE EXCLUSIVE PRICE
    192.77 EUR
    165.90 EUR / Pack of 1
    Save 26.87 EUR
    13% Off
Estimated Shipment: 05-01-2027
Highest quality Stainless Steel lid - acid proof For 608/609-150 Amsta transport container. Manufactured in Sweden.
  • Highest quality in stainless steel.
  • Acid proof 316L/1.4404.
Specifications
Product Type Container Lid
Capacity (Metric) None
Color Silver
Material Stainless Steel