Learn More
Hamilton™ Stainless Steel Large Hub Removable Custom Needles
Product Code. 12326459
Change view
Click to view available options
Needle Gauge:
22 G
22s G
23 G
23s G
25s G
26 G
26s G
Needle Point Style:
2, 3, or 4
5 or AS
Unit Size:
Each
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Needle Point Style Unit Size
12326459 25s G 2, 3, or 4 Pack of 6
10146383 22 G 5 or AS Pack of 6
11542525 22s G 2, 3, or 4 Pack of 6
10268047 23 G 2, 3, or 4 Pack of 6
10402205 22 G 2, 3, or 4 Pack of 6
12316309 23s G 5 or AS Pack of 6
15937335 26 G 2, 3, or 4 Each
11550042 26 G 2, 3, or 4 Pack of 6
10051173 26s G 2, 3, or 4 Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12326459 Supplier Hamilton™ Supplier No. 780609
  • ONLINE EXCLUSIVE PRICE
    197.00 EUR
    180.07 EUR / Pack of 6
    Save 16.93 EUR
    8% Off
Estimated Shipment: 20-10-2026
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Can be used with removable (RN)-style syringes that are 250μL and larger.
  • Available in various gauges
  • Provided in packs of 6
  • Custom point style and lengths
  • Autoclavable
  • For large hubs
Specifica
Autoclavable Autoclavable
Material Stainless Steel
Needle Gauge 25s G
Diameter (Metric) Inner 0.15 mm
Diameter (Metric) Outer 0.52 mm
Product Type Removable Custom Needle
Quantity 6/Pk.
Needle Point Style 2, 3, or 4
For Use With (Equipment) Removable Syringes (250μL and larger)
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.