Learn More
Bochem™ Stainless Steel Laboratory Scissors
Product Code. 10223941
Change view
Click to view available options
Length (Metric):
100 mm
115 mm
130 mm
145 mm
150 mm
165 mm
180 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10223941 180 mm Each
10633652 100 mm Each
10305961 115 mm Each
10214611 165 mm Each
10490923 130 mm Each
10326101 145 mm Each
10491113 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10223941 Supplier Bochem™ Supplier No. 4006
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 04-12-2026
Manufactured from stainless steel
  • Three types: round-round, sharp-round, and sharp-sharp
  • Stainless steel
Specifications
Product Type Laboratory Scissors
Length (Metric) 180 mm
Material Stainless Steel
Tip Style Sharp/Round