Learn More
Bochem™ Stainless Steel Laboratory Pot
Product Code. 10666331
Change view
Click to view available options
Capacity (Metric):
2.5 L
3.5 L
5 L
6.5 L
8.5 L
15.5 L
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10666331 2.5 L Each
10606721 3.5 L Each
10135840 5 L Each
10258320 6.5 L Each
10669852 8.5 L Each
10653072 15.5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10666331 Supplier Bochem™ Supplier No. 8360
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
Note:
Lid is not included; sold separately.
Specifications
Diameter (Metric) Inner 160 mm
Height (Metric) 128H
Capacity (Metric) 2.5 L
Material Stainless Steel (18/10)
Product Type Laboratory Pot