Learn More
Welch Ilmvac™ Stainless Steel KF Flange with Long Tubulation
Product Code. 10158121 Shop All Welch Ilmvac Products
Change view
Click to view available options
Diameter (Metric) Inner:
16 mm
24 mm
40.5 mm
Diameter (Metric) Outer:
20 mm
28 mm
44.5 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Diameter (Metric) Outer Unit Size
10158121 16 mm 20 mm Each
10551213 40.5 mm 44.5 mm Each
10433122 24 mm 28 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10158121 Supplier Welch Ilmvac™ Supplier No. 701321
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
Estimated Shipment: 04-01-2027
Specifications
Diameter (Metric) Inner 16 mm
Material Stainless Steel
Diameter (Metric) Outer 20 mm
Overall Length (Metric) 5.2 cm
Packaging Quantity 1 Each
Tubing Size DN 16
Product Type KF Flange with Long Tubulation