Learn More
Swann-Morton™ Stainless Steel Individual Foil Packs, Sterile Scalpels, SM65
Can be used in a number of specialist disciplines.
Brand:  Swann-Morton™ 5905
Additional Details : Weight : 0.02160kg
Product Code. 15337576
  • 40.55 EUR / Pack of 25
Estimated Shipment: 27-05-2024
Description
  • Versatile.
  • Can be used in a number of disciplines.
    • Plastic and reconstructive surgery
    • Orthopedics
    • Cardiovascular surgery
    • Dental periodontics
  • Fits handles SF1, SF2, SF3, SF4, SF13, and SF23.
Specifications
Specifications
Stainless Steel
Sterile
25/Box
Individually Packaged