Learn More
Usbeck Laborgeraete™ Stainless Steel Forceps
Product Code. 10005601
Change view
Click to view available options
Length (Metric):
105 mm
115 mm
130 mm
145 mm
160 mm
200 mm
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10005601 130 mm Each
10543952 105 mm Each
10790883 115 mm Each
10639522 105 mm Each
10690093 115 mm Each
10360942 130 mm Each
10005611 145 mm Each
10651452 160 mm Each
10176911 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10005601 Supplier Usbeck Laborgeraete™ Supplier No. 3012
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 23-11-2026
Specifications
Product Type Forceps
Length (Metric) 130 mm
Material Stainless Steel
Dimensions (L x W) 130 L mm
No. per Pack 1 Pack
Tip Style Blunt