Learn More
Agencinox™ Stainless Steel Footstool
Product Code. 11744926
Change view
Click to view available options
Height (Metric):
180 mm
210 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
11744926 180 mm Each
11709885 210 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11744926 Supplier Agencinox™ Supplier No. 779
  • ONLINE EXCLUSIVE PRICE
    510.30 EUR
    288.53 EUR / Pack of 1
    Save 221.77 EUR
    43% Off
Estimated Shipment: 24-11-2026
Use this footstool to safely retrieve items from high places. Agencinox™ Stainless Steel Footstools feature non-slip steps and are designed for use in laboratories and medical environments.
  • Anti-skidding steps
  • 18/10 Stainless steel construction
  • Available in a variety of sizes
Specifications
Height (Metric) 180 mm
Quantity 1 Each
Width (Metric) 340 mm
Material Stainless Steel
Product Type Foot Stool