Learn More
Agencinox™ Stainless Steel Foot Stool
Product Code. 11725635
Change view
Click to view available options
Length (Metric):
235 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11725635 235 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11725635 Supplier Agencinox™ Supplier No. 769
  • ONLINE EXCLUSIVE PRICE
    280.35 EUR
    158.67 EUR / Pack of 1
    Save 121.68 EUR
    43% Off
Estimated Shipment: 01-12-2026
Use this footstool to safely retrieve items from high places. Agencinox™ Stainless Steel Foot Stools feature non-slip steps and are designed for use in laboratories and medical environments.
  • Anti-skidding steps
  • 18/10 Stainless steel construction
  • Available in a variety of sizes
Specifications
Height (Metric) 370 mm
Length (Metric) 235 mm
Model 2 Steps
Quantity 1 Each
Width (Metric) 340 mm
Load Capacity (Metric) 100 kg
Material Stainless Steel
Product Type Foot Stool