Learn More
Bochem™ Thermo Scientific™ Stainless Steel Dressing Scissors
Product Code. 10787191
Change view
Click to view available options
Length (Metric):
130 mm
140 mm
155 mm
Tip Style:
Round/Round
Sharp/Round
Sharp/Sharp
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Tip Style Unit Size
10787191 155 mm Sharp/Round Each
10540042 130 mm Round/Round Each
10088360 130 mm Sharp/Sharp Each
10208930 140 mm Sharp/Round Each
10231961 155 mm Round/Round Each
10733521 140 mm Round/Round Each
10777191 155 mm Sharp/Sharp Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10787191 Supplier Bochem™ Supplier No. 4102
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
Manufactured from stainless steel. Bochem™ Thermo Scientific™ Stainless Steel Dressing Scissors comes in three types.
  • Three types: round-round, sharp-round, and sharp-sharp
  • Stainless steel
Specifications
Product Type Dressing Scissors
Length (Metric) 155 mm
Material Stainless Steel
Tip Style Sharp/Round