Learn More
Fisherbrand™ Stainless Steel Cylindrical Scoops
Product Code. 11553452 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
190 mm
260 mm
325 mm
340 mm
Unit Size:
Each
Capacity (Metric):
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize Capacity (Metric)
11553452 190 mm Each -
11563452 260 mm Each -
11573452 325 mm Each 500 mL
11583452 340 mm Each 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11553452
  • ONLINE EXCLUSIVE PRICE
    22.00 EUR
    18.56 EUR / Pack of 1
    Save 3.44 EUR
    15% Off
View all promotions
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Get optimum results weighing and handling samples and other laboratory materials with cylindrical scoops. Perfect for weighing anything from liquid to gems and other materials, Fisherbrand™ Stainless Steel Cylindrical Scoops are an ideal accessory to have if you own a weighing scale.
  • Manufactured from stainless steel.
  • Cylindrical shape keeps materials from spilling
  • Includes handle
Specifications
Shape Cylindrical
Product Type Scoop
Dimensions (L x W) 190 x 50 mm
Includes With Handle
Material Stainless Steel
Length (Metric) 190 mm