Learn More
Fisherbrand™ Stainless Steel Round Bottom Weighing Scoops
Product Code. 11533452 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
70 mm
90 mm
105 mm
120 mm
Unit Size:
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11533452 105 mm Pack of 5
11543452 120 mm Pack of 5
11513452 70 mm Pack of 5
11523452 90 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11533452
Only null left
Add to basket
Add to basket
Get optimum results weighing and handling samples and other laboratory materials with round bottom scoops. Perfect for weighing anything from liquid to gems and other materials, Fisherbrand™ Stainless Steel Round Bottom Weighing Scoops are an ideal accessory to have if you own a weighing scale.
  • Well-suited for pharmaceutical and environmental sampling
  • Ideal for weighing and handling of samples
  • Great for weighing messy ingredients, tiny objects and portions
  • Great accessory to have if you own a weighing scale
  • Perfect for weighing anything from liquid to gems and other materials
Specifications
Shape Cylindrical
Product Type Scoop
Width (Metric) 30 mm
Dimensions (L x W) 105 x 30 mm
Material Stainless Steel
Length (Metric) 105 mm