Learn More
IKA™ Stainless Steel Cutter
Shop All IKA Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Description
- Stainless steel construction
- For use with batch mills
- For cutting, grinding, or pulverizing soft, fibrous materials with a Mohs hardness up to 6
Specifications
Specifications
| Material | Stainless Steel |
| Packaging Quantity | 1 Ea. |
| Weight (Metric) | 0.007 kg |
Product Content Correction
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu